Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70091-M

Sigma-Aldrich

pET-40b(+) DNA - Novagen

Synonym(s):

pET Dsb Fusion System 39b and 40b, pET System, pET- 40b(+) vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.51

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice
wet ice

storage temp.

−70°C

General description

The pET Dsb Fusion Systems 39b and 40b are designed for cloning and high-level expression of peptide sequences fused with the 208-aa DsbA-Tag sequence [pET-39b(+)] or the 236-aa DsbC-Tag sequence [pET-40b(+)]. DsbA and DsbC are periplasmic enzymes that catalyze the formation and isomerization of disulfide bonds, respectively (Rietsch 1996, Sone 1997, Missiakas 1994, Zapun 195, Raina 1997). In contrast to the pET-32 Trx-Tag series, which is designed to increase solubility in the cytoplasm, the DsbA-Tag and DsbC-Tag vectors enable potential periplasmic localization and thus may enhance solubility and proper folding in this non-reducing environment (Collins-Racie 1995).

he pET-40b(+) vector is designed for expression of DsbC fusion proteins. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map (TB229). The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. The pET Vectors are supplied as purified plasmid DNA (10 µg). Each order of pET DNA also includes an Induction Control strain (supplied as a glycerol stock). Please contact technical service if you need additional information.

Warning

Toxicity: Standard Handling

Other Notes

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the ′EMD Product′) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service