Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB8123

Sigma-Aldrich

Anti-phospho-Integrin β1 (Thr788/789) Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr788/pThr789)

Gene Information

human ... ITGB1(3688)

Specificity

Integrin B1 phosphorylated on Thr788/789

Immunogen

C-terminal Peptide

Application

Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-phospho-Integrin β1 (Thr788/789) Antibody is validated for use in WB for the detection of phospho-Integrin β1 (Thr788/789).

Quality

Routinely evaluated by immunoblot.

Target description

120kDa

Physical form

Raw Rabbit Serum with 0.1% Sodium Azide

Storage and Stability

1 year at 4°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service