Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF1809

Sigma-Aldrich

Mouse IgG1-k Negative Control, clone MOPC-21, PerCP conjugate Antibody

clone MOPC-21, from mouse

Synonym(s):

Immunoglobulin G1, IgG1-k

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

antibody form

purified antibody

clone

MOPC-21, monoclonal

technique(s)

flow cytometry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
The isotype control should be used at the same concentration as the primary antibody for which it is a control.
This mouse monoclonal Mouse IgG1-k Negative Control, clone MOPC-21, PerCP conjugate, Cat. No. MABF1809 is validated for use in Flow Cytometry, for the detection of IgG1-k.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography.

Physical form

Format: Purified
Purified Mouse monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service