Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF2685

Sigma-Aldrich

Anti-Influenza A HA Antibody, clone rFluA-20 Antibody, Recombinant Monoclonal

clone rFluA-20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

clone

rFluA-20, recombinant monoclonal

Quality Level

isotype

IgG1

Specificity

Clone rFluA-20 is human recombinant monoclonal antibody that detects Influenza A virus hemagglutinin in multiple strains.

Immunogen

PBMCs from human donor, who was subjected to Flu vaccine multiple times, were immortalized and used generate human hybridoma cell lines secreting human mAbs.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service