MABF2698
Anti-P. aeruginosa serogroup It-5 Antibody, clone PTA-5385
Synonym(s):
Pseudomonas aeruginosa It-5 serotype
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
PTA-5385, monoclonal
mol wt
calculated mol wt N/A kDa
observed mol wt ~N/A kDa
purified by
using protein G
species reactivity
bacteria
packaging
antibody small pack of 100
technique(s)
ELISA: suitable
inhibition assay: suitable
isotype
IgG2κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Specificity
Immunogen
Application
Evaluated by ELISA with LPS specific for serogroup It-5.
ELISA Analysis: Serial dilutions of this antibody detected LPS from Pseudomonas aeruginosa serogroup It-5.
Tested Applications
ELISA Analysis: A representative lot detected LPS from Pseudomonas aeruginosa (It-5) in ELISA applications (Lai, Z., et al. (2005). Vaccine. 23(25):3264-71).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service