Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

MABF2784

Sigma-Aldrich

Anti-SARS-CoV-2 Spike RBD Antibody, clone 6E4-C5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies, primary antibodies

clone

6E4-C5, monoclonal

mol wt

calculated mol wt 141.18 kDa
observed mol wt ~200 kDa

purified by

using protein G

species reactivity

SARS coronavirus

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

vaccinia virus ... NS1(956533)

General description

Spike glycoprotein (UniProt: P0DTC2; also known as S glycoprotein, E2, Peplomer protein) is encoded by the S (also known as 2) gene (Gene ID: 43740568) in Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2). The SARS-CoV-2 is a positive-strand RNA virus that causes severe respiratory syndrome in human. The mature SARS-CoV-2 contains 4 structural proteins: Envelope (E), Membrane (M), Nucleocapsid (N), and the Spike protein (S). E and M proteins help in viral assembly and N protein is needed for RNA synthesis. The S protein is a single-pass type I, homotrimeric, membrane glycoprotein that is responsible for virus binding and entry into host cell. It is synthesized with a signal peptide (aa 1-12), which is subsequently cleaved off to generate the mature protein that contains an extracellular domain (aa 13-1213), a transmembrane domain (aa 1214-1234), and a cytoplasmic domain 1235-1273). The S protein is further processed into S1 (aa 13-685) and S2 (aa 686-1273) subunits by host cell furin. The presence of a furin polybasic cleavage site in S protein from SARS-CoV-2 sets it apart from S protein in SARS-CoV that possesses a monobasic S1/S2 cleavage site. The S1 subunit has the receptor binding domain (RBD; aa 319-541) that mediates entry of SARS-CoV-2 into sensitive cells through the peptidase domain of host Angiotensin-converting enzyme 2 (ACE2) with high affinity (KD = 15 nM). The S2 domain (aa 686-1213), which is reported to be well conserved, is responsible for membrane fusion. (Ref.: Abe, KT., et al. (2020). JCI Insight. 5(19); e142362).

Specificity

Clone 6E4-C5 is a mouse monoclonal antibody that detects the spike glycoprotein of SARS-CoV-2. It targets an epitope within the receptor binding domain.

Immunogen

His-tagged recombinant fragment corresponding to 223 amino acids from receptor binding domain of SARS-CoV-2.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HEK293T cells expressing full-length spike protein of SARS-CoV-2.

Western Blotting Analysis: A 1:500 dilution of this antibody detected spike glycoprotein in lysate from HEK293T cells expressing full-length SARS-CoV-2 spike protein, but not in lysate from wild type HEK393T cells.

Tested Applications

Neutralizing: A representative lot of this antibody reduced binding of SARS-CoV-2 RBD to ACE2 receptor in a Binding Interference Assay. (Courtesy: Kento Abe and Anne-Claude Gingras, University of Toronto, Canada; Abe, KT., et al. (2020). JCI Insight. 5(19); e142362).

ELISA Analysis: Various dilutions of a hybridoma culture supernatant for clone 6E4-C5 detected native SARS-CoV-2 spike protein. ( (Courtesy of Stefan Schüchner, Ingrid Mudrak, Ingrid Frohner, and E. Ogris (Max Perutz Labs, Medical University of Vienna, Austria).

Immunoprecipitation Analysis: A representative lot immunoprecipitated spike glycoprotein of SARS-CoV-2. (Courtesy of Stefan Schüchner, Ingrid Mudrak, Ingrid Frohner, and E. Ogris (Max Perutz Labs, Medical University of Vienna, Austria).

Western Blotting Analysis: A 1:400 dilution from a representative lot detected spike glycoprotein in whole cell lysate of HEK293T cells expressing full-length spike protein of SARS-CoV-2, but not in untransfected cells. (Courtesy of Stefan Schüchner, Ingrid Mudrak, Ingrid Frohner, and E. Ogris (Max Perutz Labs, Medical University of Vienna, Austria).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SARS-CoV-2 Spike RBD, clone 6E4-C5, Cat. No. MABF2784, is a mouse monoclonal antibody that detects spike glycoprotein of SARS-CoV-2 and is tested for use in ELISA, Immunoprecipitation, Neutralizing, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service