Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2296

Sigma-Aldrich

Anti-C9orf72 peptide 4 Antibody, clone 12E7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

12E7, monoclonal

mol wt

calculated mol wt 54.33 kDa
observed mol wt ~54 kDa

purified by

using protein G

species reactivity

mouse, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... C9orf72(203228)

General description

Guanine nucleotide exchange C9orf72 (UniProt: Q96LT7; also known as C9orf72) is encoded by the C9orf72 gene (Gene ID: 203228) in human. C9orf72 generates three transcripts through alternative splicing that encode 2 protein isoforms: a long isoform of about 54 kDa (termed C9-L), corresponding to variants 2 and 3, and a short isoform of about 24 kDa (termed C9-S) corresponding to variant 1. Both isoforms are widely expressed, including in kidney, lung, liver, heart, testis, and several brain regions. The long isoform of 481 amino acids is shown to be the predominant form that is expressed in human brain. Expansion of a GGGGCC (G4C2) hexanucleotide repeat sequence in the non-coding region of human C9orf72 is the most common genetic abnormality in familial and sporadic frontotemporal dementia (FTD) and motor neuron disease (MND), with amyotrophic lateral sclerosis (ALS) as the most frequent form. Both, C9-L and C9-S are reported to display distinct subcellular localizations in cerebellar Purkinje cells of both c9-ALS and non c9-ALS cases. C9-L is shown to be spread diffusely throughout the cytoplasm, whereas C9- S is localized to the nuclear membrane of Purkinje cells of both c9-ALS and non-c9-ALS cases. Also, C9-L is shown to be present mainly in the high-salt Triton and urea-soluble fractions, whereas C9-S is shown to be abundant in both the low- and high-salt Triton fractions, indicating differences in their subcellular localizations. (Ref.: Frick, P., et al. (2018). Acta Neuropathol. Commun. 6(1); 72).

Specificity

Clone 12E7 is a rat monoclonal antibody that detects Guanine nucleotide exchange C9orf72. It targets an epitope within 16 amino acids from the N-terminal half.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of human Guanine nucleotide exchange C9orf72.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:250 dilution of this antibody detected C9orf72 in HEK293 cell lysate.

Tested Applications

Immunohistochemistry Applications: A representative lot detected C9orf72 in Immunohistochemistry applications (Frick, P., et al. (2018). Acta Neuropathol Commun. 6(1):72).

Western Blotting Analysis: A representative lot detected C9orf72 in Western Blotting applications (Frick, P., et al. (2018). Acta Neuropathol Commun. 6(1):72).

Immunofluorescence Analysis: A representative lot detected C9orf72 peptide 4 in Immunofluorescence applications (Frick, P., et al. (2018). Acta Neuropathol Commun. 6(1):72).

Immunocytochemistry Analysis: A representative lot detected C9orf72 peptide 4 in Immunocytochemistry applications (Frick, P., et al. (2018). Acta Neuropathol Commun. 6(1):72).

ELISA Analysis: A representative lot detected C9orf72 peptide 4 in ELISA applications (Frick, P., et al. (2018). Acta Neuropathol Commun. 6(1):72).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-C9orf72 peptide 4, clone 12E7, Cat. No. MABN2296, is a rat monoclonal antibody that detects C9orf72 and is tested for use in ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service