Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2189

Sigma-Aldrich

Anti-PTPRD Antibody, clone F34a6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

F34a6
monoclonal

purified by

using protein G

species reactivity

mouse, mouse, rat

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

General description

GST-tagged recombinant fragments corresponding to Fibronectin type III repeats 2,3 and 8 of murine Receptor-type tyrosine-protein phosphatase delta (PTPRD).

Specificity

Clone F34a6 is a rat monoclonal antibody that specifically detects different isoforms of murine Receptor-type tyrosine-protein phosphatase delta (PTPRD), but does not react with LAR. It targets an epitope within the ectodomain region.

Application

Epitope: extracellular domain

Quality

Evaluated by Western Blotting in E16 mouse brain tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected receptor-type tyrosine-protein phosphatase delta (PTPRD) in E16 mouse brain tissue lysate.

Physical properties

~80, 120, 130 kDa observed, 214.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service