Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PR0GMPK01

Milli-Q

Kit: Progard® 1 Pack & Vent Filter

Protects the RO membrane and preserves stored water (Type 2) quality

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
NB.85

Quality Level

feature

Kit contains Progard® 1 Pretreatment Pack and a Tank Vent Filter for Type 2 water

packaging

pkg of 1 kit

compatibility

for use with Direct-Q®
for use with Elix®
for use with RiOs

General description

This consumable kit includes Progard® Pretreatment Pack and a Tank Vent Filter.
  • Pretreatment Pack is designed to pretreat feed water by removing large particles, organic compounds, and free chlorine from tap water. This protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling and chlorine oxidation.

Tank Vent Filter prevents airborne contaminants from entering the pure water storage tank.

Application

For optimal protection of the RO membrane and pure water in the storage tank.

Features and Benefits

  • Optimal protection of RO membrane.
  • Protects the quality of stored pure water.
  • Ensures prolonged system lifetime and optimizes performance.

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service