Recommended Products
description
Cell line type: Neural Stem Cells
Neural Stem Cells
Quality Level
technique(s)
cell culture | stem cell: suitable
immunocytochemistry: suitable
General description
The Adult Rat Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies. This system includes primary neural stem cells isolated from the hippocampus of adult Fisher 344 rats, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes).
Components
Adult Rat Hippocampal Neural Stem Cells (Cat.# SCR022) Rat Neural Stem Cell Expansion Medium (Cat.# SCM009) Neural Stem Cell Marker Characterization Kit (Cat.# SCR019)
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service