Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3290

Dry Cat Food

NIST® SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

format

matrix material

General description

A unit of SRM 3290 consists of five heat sealed, aluminized pouches, each containing approximately 10 g of blended, commercially available dry cat foods.SRM 3290_cert SRM 3290 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service