Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR8525

Supelco

NMBA-D3

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

d3-4-[Methyl(nitroso)amino]butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7D3N2O3
CAS Number:
Molecular Weight:
149.16
UNSPSC Code:
12352200

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

(Not traceable to any primary pharmacopeia standard)

CofA

current certificate can be downloaded

packaging

pkg of 1.2 mL

concentration

1 mg/mL in acetonitrile

application(s)

pharmaceutical

storage temp.

-10 to -25°C

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

Stable Isotoped Labelled product intended for use as internal standard with mass spectrometry detection.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service