Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00957

Sigma-Aldrich

Vanylglycol sodium salt

≥95% (HPLC)

Synonym(s):

4-Hydroxy-3-methoxy-β-phenylglycol sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NaO4
Molecular Weight:
206.17
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white to brown

shipped in

wet ice

storage temp.

2-8°C

Application

Vanylglycol is also known as MHPG or MOPEG and can be biosynthesized from 3,4-dihydroxyphenylglycol and guaiacol. MHPG is a O-methylated metabolite of normetanephrine. Alcohol consumption increases the level of Vanylglycol in urine and CSF. Vanylglycol is found normally in urine, in plasma and cerebrospinal fluid. Outside of the human body, vanylglycol has been detected, but not quantified in several different foods such as blackcurrants, chinese bayberries, elderberries, oriental wheats, and poppies. This could make vanylglycol a potential biomarker for the consumption of these foods. Vanylglycol has been found to be associated with several diseases such as multi-infarct dementia, epilepsy, hereditary spastic paraplegia, and schizophrenia; also vanylglycol has been linked to the inborn metabolic disorders including aromatic l-amino acid decarboxylase deficiency.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service