Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VT002132

Millipore

Cronobacter muytjensii WDCM 00213 Vitroids

greener alternative

15-80 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

sustainability

Greener Alternative Product

application(s)

microbiology

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of our four categories of greener alternatives. Click here to view its DfS scorecard.

Legal Information

Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service