Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OX-90

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Mouse Hybridoma

packaging

tube of 5 μg 03062502-DNA-5UG
pkg of vial of cells 03062502-1VL

ImmunoDon

PVG rat

Myeloma

NS1

Reactivity

Anti-CD200 (OX2), mouse

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS1 x rat

Cell Line Description

This antibody recognises the CD200 antigen present on a variety of cells including activated T lymphocytes, neurons, follicular dendritic cells and endothelium.

Immunogen

Recombinant mOX-2-rCD4 d3-4

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.

Culture Medium

RPMI 1640 +2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Seed at 5 x 10,000 cells/ml. Subculture when cells reach 500,000 - 1,000,000 cells/ml (usually after 3-4 days), 5% CO2, 37°C. Start in 20% FBS when recovering from frozen.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service