Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JW8/5/13

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl), 87080706

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse + Human Transfectoma

packaging

tube of 5 μg 87080706-DNA-5UG
vial of  cells 87080706-1VL

ImmunoDon

Not applicable

Myeloma

J558L

Reactivity

Anti-NP (4-hydroxy-3 nitrophenacetyl)

technique(s)

cell culture | mammalian: suitable

isotype

IgE

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse J558L + Mouse VH gene + Human C (epsilon) gene

Cell Line Description

This human chimeric antibody is directed against the hapten 4-hydroxy-3 nitrophenacetyl. The V lambda of the endogenous light chain of the plasmacytoma line complements the VNP of the transfected heavy chain to yield the specific antibody. Gene marker: gpt.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service