Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MOPC 31C

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 90110707, mouse unknown/unspecified, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse unknown/unspecified

packaging

tube of 5 μg 90110707-DNA-5UG
pkg of vial of cells 90110707-1VL

growth mode

Suspension

karyotype

Modal no. 43, 81-85 hyperdiploid

morphology

Lymphoblast

products

Immunoglobulin G (IgG1)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c plasmacytoma, IgG secreting

Cell Line Description

The original plasma cell tumour was induced by intraperitoneal injections of mineral oil into highly inbred BALB/cJ mice. The line secretes IgG1, is hyperdiploid and produces a balanced synthesis of monomeric 4-chain gamma F/immunoglobulin.

Application

Tumourigenicity studies

Culture Medium

L15 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 37°C. The cells have a maximum density of 1-2x1,000,000 cells/ml. CO2 is not required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service