Key Documents
MTCV5920
MTC Bio Locking Rack for Cryogentic Vials
to hold, 5 x 10 cryovials
Synonym(s):
flask screw cap, vial holder, vial storage
About This Item
UNSPSC Code:
41122806
NACRES:
NB.12
material
polypropylene
feature
to hold 5 x 10 cryovials
packaging
pack of 4 ea
manufacturer/tradename
MTC Bio V5920
General description
Polypropylene rack for securely holding 50 "star foot" vials, 210 × 100 × 25mm, so that one hand is all that is needed to install and remove caps.
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)