SAB3501150
Anti-SARS-CoV-2 (COVID-19) Spike (cleavage site) (biotin) antibody produced in rabbit
affinity isolated antibody
biological source
rabbit
Quality Level
conjugate
biotin conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
species reactivity
SARS coronavirus
concentration
1 mg/mL
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
SARS coronavirus ... S(43740568)
General description
Immunogen
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service