Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4702109

Monoclonal Anti-CD41/CD61 (PAC-1 epitope) FITC produced in mouse

new

Synonym(s):

GP3A, GPIIb, GPalpha IIb, GT, Integrin alpha-Iib, platelet GPIIIa, platelet GPIIb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

FITC conjugate

Quality Level

antibody product type

primary antibodies

clone

PAC-1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgMκ

storage temp.

2-8°C

target post-translational modification

unmodified

General description

CD41 (platelet glycoprotein IIb, integrin alpha IIb) is composed of two subunits (120 kDa transmembrane alpha chain and 23 kDa extracellular beta chain) and interacts with CD61 (platelet glycoprotein IIIa, integrin beta 3) in the presence of calcium to form a functional adhesive protein receptor. CD41/CD61 complex is one of the earliest markers of the megakaryocytic lineage. Upon blood vessel damage, this receptor binds to a variety of proteins including von Willebrand factor, fibrinogen, fibronectin and vitronectin, and it is involved in platelet aggregation.

Specificity

The mouse monoclonal antibody PAC-1 recognizes an extracellular activation-induced conformational epitope PAC-1 on CD41/CD61 complex (gpIIb/IIIa), also known as integrin alpha IIb beta 3, a receptor which mediates platelet aggregation.

Immunogen

Human platelets

Physical form

Stabilizing Tris buffered saline (TBS), pH 8.0, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with fluorescein isothiocyanate (FITC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service