Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4702151

Monoclonal Anti-CD300e, purified low endotoxin produced in mouse

new

Synonym(s):

CLM2, CMRF35-A5, IREM-2, LMIR6, PlgR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

UP-H2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

FUNC: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD300E(342510)

General description

CD300e / IREM-2 (immune receptor expressed by myeloid cells 2), also known as CLM2 or LMIR6, is a monomeric transmembrane glycoprotein with a single extracellular immunoglobulin-like domain. Intracellularly it associates with DAP-12, an ITAM-containing adaptor molecule. CD300e is expressed on mature monocytes and peripheral blood myeloid dendritic cells. Its crosslinking leads to release of pro-inflammatory cytokines, and increased expression of activation markers.

Specificity

The mouse monoclonal antibody UP-H2 recognizes an extracellular epitope on CD300e / IREM-2, a 32 kDa glycoprotein expressed by mature monocytes and peripheral blood myeloid dendritic cells.

Immunogen

CD300e-HA-transfected cells

Physical form

Phosphate buffered saline (PBS), pH 7.4

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service