Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCP0160

Sigma-Aldrich

GRF, mouse

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C220H365N69O64S1
Molecular Weight:
5032.74
UNSPSC Code:
51111800
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

His-Val-Asp-Ala-Ile-Phe-Thr-Thr-Asn-Tyr-Arg-Lys-Leu-Leu-Ser-Gln-Leu-Tyr-Ala-Arg-Lys-Val-Ile-Gln-Asp-Ile-Met-Asn-Lys-Gln-Gly-Glu-Arg-Ile-Gln-Glu-Gln-Arg-Ala-Arg-Leu-Ser

Application

Growth Hormone Releasing Factor (GHRF) is an amino acid peptide that induces the release of growth hormone via GPCR receptor GHRH-R.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service