Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2962

Sigma-Aldrich

MM07 trifluoroacetate

≥94.5% (HPLC)

Synonym(s):

Cys-Arg-Pro-Arg-Leu-Cys-His-Lys-Gly-Pro-Met-Pro-Phe disulfide bridge 1-6 trifluoroacetate, L-cysteinyl-L-arginyl-L-prolyl-L-arginyl-L-leucyl-L-cysteinyl-L-histidyl-L-lysyl-glycyl-L-prolyl-L-methionyl-L-prolyl-L-phenylalanine (1->6)-disulfide trifluoroacetate, cyclo[1-6]CRPRLCHKGPMPF trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C67H106N22O14S3
Molecular Weight:
1539.89
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥94.5% (HPLC)

form

lyophilized powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

G protein biased apelin receptor agonist
MM07 is a G protein biased apelin receptor agonist that causes rapid and significant peripheral arterial dilatation in human hand vein. MM07 is more stable than the endogenous apelin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service