Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3114

Sigma-Aldrich

R121919

≥98% (HPLC)

Synonym(s):

3-[6-(Dimethylamino)-4-methyl-3-pyridinyl]-2,5-dimethyl-N,N-dipropylpyrazolo[1,5-a]pyrimidin-7-amine, NBI 30775, NBI-30775, NBI30775, R 121919, R-121919

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32N6
CAS Number:
Molecular Weight:
380.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCN(CCC)C1=CC(C)=NC2=C(C(C)=NN12)C3=CN=C(C=C3C)N(C)C

InChI

1S/C22H32N6/c1-8-10-27(11-9-2)20-13-16(4)24-22-21(17(5)25-28(20)22)18-14-23-19(26(6)7)12-15(18)3/h12-14H,8-11H2,1-7H3

InChI key

ANNRUWYFVIGKHA-UHFFFAOYSA-N

Biochem/physiol Actions

Brain-penetrant and orally available selective CRF1 (CRFR-1, CRHR-1) antagonist with in vivo antistress efficacy.
R121919 (NBI 30775) is a brain-penetrant, orally available high-affinity corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1) antagonist (Ki = 3.5 nM against sauvagine for binding hCRFR1-expressing HEK293 cells; IC50 = 20 nM/CRF-stimulated ACTH release from rat anterior pituitary cells & IC50 = 80 nM/CRF-stimulated cAMP production in hCRFR1-expressing HEK293 cells) without affinity toward CRFR2. In addition to its antistress efficacy in various rat anxiety models (0.63-20 mg/kg p.o.), R121919 also demonstrates therapeutic efficacy and long-term safety in Alzheimer′s disease (AD) mice (20 mg/kg/day s.c.) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service