Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3130

Sigma-Aldrich

SHetA2

≥98% (HPLC)

Synonym(s):

1-(4-Nitrophenyl)-3-(2,2,4,4-tetramethylthiochroman-6-yl)thiourea, N-(3,4-Dihydro-2,2,4,4-tetramethyl-2H-1-benzothiopyran-6-yl)-N′-(4-nitrophenyl)thiourea, NSC 726189, NSC-726189, NSC726189, Sulfur Heteroarotinoid A2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23N3O2S2
CAS Number:
Molecular Weight:
401.55
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C(NC1=CC=C2SC(C)(CC(C)(C)C2=C1)C)NC(C=C3)=CC=C3[N+]([O-])=O

InChI

1S/C20H23N3O2S2/c1-19(2)12-20(3,4)27-17-10-7-14(11-16(17)19)22-18(26)21-13-5-8-15(9-6-13)23(24)25/h5-11H,12H2,1-4H3,(H2,21,22,26)

InChI key

FWWKIYPMUZVMQG-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available chemopreventive fexible heteroarotinoid (Flex-Het) that induces cancer apoptosis in cultures and in vivo independent of retinoic acid receptor (RAR).
SHetA2 is an orally available chemopreventive fexible heteroarotinoid (Flex-Het) that inhibits cancer growth in cultures (OVCAR-3/SK-OV-3/Caov-3 GI50 = 3.9/4.1/7.0 μM) and in vivo (60-100 mg/kg SiHa xenograft mice) via apoptosis induction independent of retinoic acid receptor (RAR). SHetA2 is reported to bind heatshock proteins HSPA5 (GRP78, BiP), HSPA8 (HSC70, HSP73, HSPA10), HSPA9 (GRP75), resulting in cyclin D1 degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service