Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3299

Sigma-Aldrich

S-Ketorolac

≥98% (HPLC)

Synonym(s):

(-)-Ketorolac, (1S)-5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid, (S)-Ketorolac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO3
CAS Number:
Molecular Weight:
255.27
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -165 to -185°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]21c(ccc2C(=O)c3ccccc3)[C@H](CC1)C(=O)O

InChI

1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/t11-/m0/s1

InChI key

OZWKMVRBQXNZKK-NSHDSACASA-N

Biochem/physiol Actions

S-Ketorolac is an active enantiomer of nonsteroidal anti-inflammatory drug Ketorolac. S-Ketorolac is a selective and potent cyclooxygenase 1/2 (COX-1/2) inhibitor which exhibits anti-inflammatory activities. It is significantly more potent than non-selective COX inhibitors indomethacin or diclofenac.
selective and potent COX-1/2 inhibitor

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service