Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3440

Sigma-Aldrich

AVN-944

≥98% (HPLC)

Synonym(s):

(1R)-1-(Cyanomethyl)propyl N-[(1S)-1-[3-[[[[3-methoxy-4-(5-oxazolyl)phenyl]amino]carbonyl]amino]phenyl]ethyl]carbamate, AVN 944, VX-944, [(1S)-1-[3-[[[[3-Methoxy-4-(5-oxazolyl)phenyl]amino]carbonyl]amino]phenyl]ethyl]-carbamic acid (1R)-1-(cyanomethyl)propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27N5O5
CAS Number:
Molecular Weight:
477.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N([C@@H](C)c1cc(ccc1)NC(=O)Nc2cc(c(cc2)c3[o]cnc3)OC)C(=O)O[C@H](CC)CC#N

InChI key

GYCPCOJTCINIFZ-OXJNMPFZSA-N

Biochem/physiol Actions

AVN-944 is an orally bioavailable, highly selective, nonnucleoside and uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with antitumor and antiviral activities. AVN-944 depletes intracellular GMP levels and inhibits DNA and RNA viruses replication via reduction of GTP pools in infected cells. Arrests a wide range of hematologic and epithelial tumor cell types proliferation (IC50 of 0.02 µM to 0.279 µM).
Orally bioavailable, highly selective, nonnucleoside and uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with antitumor and antiviral activities

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service