Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3479

Sigma-Aldrich

Hydroxy-α-sanshool

≥95% (HPLC)

Synonym(s):

(2E,6Z,8E,10E)-N-(2-Hydroxy-2-methylpropyl)-2,6,8,10-dodecatetraenamide, 2E,6Z,8E,10E-Dodecatetraenoic acid N-(2-hydroxy-2-methylpropyl)amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO2
CAS Number:
Molecular Weight:
263.38
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light
under inert gas

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N(CC(O)(C)C)C(=O)\C=C\CC\C=C/C=C/C=C/C

InChI

1S/C16H25NO2/c1-4-5-6-7-8-9-10-11-12-13-15(18)17-14-16(2,3)19/h4-9,12-13,19H,10-11,14H2,1-3H3,(H,17,18)/b5-4+,7-6+,9-8-,13-12+

InChI key

LHFKHAVGGJJQFF-UEOYEZOQSA-N

Biochem/physiol Actions

Hydroxy-α-sanshool is an active ingredient in Szechuan peppers (also present in other Zanthoxylum plants) that produces a tingling paresthesia or numbing effects. Hydroxy-α-sanshool is a potent agonist of TRPV1/TRPA1 as well as antagonist/blocker of KCNK3/KCNK9/KCNK18 channels. It induces colonic motor activity in rat proximal colon.
Potent agonist of TRPV1/TRPA1 as well as antagonist/blocker of KCNK3/KCNK9/KCNK18 channels

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service