Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3659

Sigma-Aldrich

SJB2-043

≥98% (HPLC)

Synonym(s):

2-Phenyl-naphth[2,3-d]oxazole-4,9-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H9NO3
CAS Number:
Molecular Weight:
275.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Faint green to yellow

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

n1c2c([o]c1c4ccccc4)C(=O)c3c(cccc3)C2=O

InChI

1S/C17H9NO3/c19-14-11-8-4-5-9-12(11)15(20)16-13(14)18-17(21-16)10-6-2-1-3-7-10/h1-9H

InChI key

CMYQQADDUUDCCA-UHFFFAOYSA-N

Biochem/physiol Actions

SJB2-043 is a ubiquitin-specific protease USP1 inhibitor (IC50 = 544 nM using USP1/UAF1 complex) that exhibits anti-proliferation efficacy against chronic and acute myeloid leukemia lines (IC50 <2 µM; K562, OCI-AML3, SK-NO1, U937) and primary acute myelogenous leukemia (AML) patient-derived leukemic cells by promoting the degradation of transcription factor DNA binding 1 (ID1). SJB2-043 is also reported to be an allosteric site-targeting inhibitor against PLPro (IC50 = 0.56 μM), the papain-like cysteine protease with deubiquitination and deISGylation activities that suppress host innate immune responses toward SARS-CoV-2 infection.
Ubiquitin-specific protease USP1 inhibitor with anti-leukemia efficacy. Also reported to inhibit SARS-CoV-2 papain-like cysteine protease PLPro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service