Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3945

Sigma-Aldrich

ARN11391

≥98% (HPLC)

Synonym(s):

2-Ethyl-N-[2-(oxan-2-ylmethoxy)phenyl]-4,5,6,7-tetrahydroindazole-3-carboxamide, ARN 11391, ARN-11391

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N3O3
CAS Number:
Molecular Weight:
383.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Selective inositol triphosphate receptor type 1 (ITPR1) potentiator for wild-type as well as two ITPR1 SCA29 mutants, R269W or T267M.

ARN11391 is a selective inositol triphosphate receptor type 1 (ITPR1), but not ITPR2 or ITPR3, potentiator (open channel probability (Po) = 0.2 with 20 µM ARN11391 and <0.01 without by on-nucleus nuclear patch-clamp using TPR1-YFP-expressing HEK293 cells) that potenitates IP3- (by photolysis of caged IP3) and UPT-induced calcium increase in HEK293 transfectants expressing wild-type as well as ITPR1 SCA29 mutants, R269W or T267M (5 µM UTP with or without 10 µM ARN11391).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service