Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4009

Sigma-Aldrich

Zuclopenthixol decanoate

≥98% (HPLC)

Synonym(s):

Decanoic acid 2-[4-[(3Z)-3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazinyl]ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H43ClN2O2S
CAS Number:
Molecular Weight:
555.21
MDL number:
UNSPSC Code:
12352116
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

oil

color

, Colorless to light brown

storage temp.

-10 to -25°C

SMILES string

ClC(C=C1)=CC2=C1SC3=CC=CC=C3/C2=C/CCN4CCN(CCOC(CCCCCCCCC)=O)CC4

InChI

1S/C32H43ClN2O2S/c1-2-3-4-5-6-7-8-15-32(36)37-24-23-35-21-19-34(20-22-35)18-11-13-27-28-12-9-10-14-30(28)38-31-17-16-26(33)25-29(27)31/h9-10,12-14,16-17,25H,2-8,11,15,18-24H2,1H3/b27-13-

InChI key

QRUAPADZILXULG-WKIKZPBSSA-N

Biochem/physiol Actions

Long-lasting injectable formulation (depot form) of Zuclopenthixol; potent antagonist of D1/D2 dopamine receptors, α1-adrenergic receptors and 5-HT2 receptors.

Zuclopenthixol decanoate is a long-lasting injectable formulation (depot form) of Zuclopenthixol. Zuclopenthixol decanoate is a potent antagonist of D1/D2 dopamine receptors, α1-adrenergic receptors and 5-HT2 receptors. Zuclopenthixol decanoate is approved in some countries for treatment of schizophrenia and other psychoses and recommended for patients who are agitated or aggressive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service