Skip to Content
MilliporeSigma

Skip To

SML4010

KPT-6566

≥98% (HPLC)

Synonym(s):

2-[[4-[[[4-(1,1-Dimethylethyl)phenyl]sulfonyl]imino]-1,4-dihydro-1-oxo-2-naphthalenyl]thio]acetic acid, KPT 6566, KPT6566, [[4-[[[4-(1,1-Dimethylethyl)phenyl]sulfonyl]imino]-1,4-dihydro-1-oxo-2-naphthalenyl]thio]acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H21NO5S2
CAS Number:
Molecular Weight:
443.54
MDL number:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(\N=C2\c3c(cccc3)C(=O)C(=C\2)SCC(=O)O)c1ccc(cc1)C(C)(C)C

InChI

1S/C22H21NO5S2/c1-22(2,3)14-8-10-15(11-9-14)30(27,28)23-18-12-19(29-13-20(24)25)21(26)17-7-5-4-6-16(17)18/h4-12H,13H2,1-3H3,(H,24,25)/b23-18+

InChI key

BXWWOKYIKNEEHJ-PTGBLXJZSA-N

Biochem/physiol Actions

Potent and selective peptidyl-prolyl cis-trans isomerase (PPIase) Pin1 covalent inhibitor with anti-cancer efficacy in cultures and in vivo.

KPT-6566 is a potent and selective peptidyl-prolyl cis-trans isomerase (PPIase) Pin1 inhibitor (IC50 = 640 nM) that targets Pin1 catalytic site for covalent modification, resulting in Pin1 degradation and the release of ROS-generating and DNA-damaging quinone-mimicking KPT-6566-B. KPT-6566 inhibits the proliferation of wild-type, but not Pin1-/-, MEFs (1-5 µM for 48h) with concomitant reduction of cellular pRB and Cyclin D1 hyperphosphorylation. KPT-6566 induces cancer cell death in cultures (1-10 µM for 48h) and reduces MDA-MB-231 lung metastasis in mice in vivo (5 mg/kg/d i.p.). KPT-6566 is more potent than juglone and does not affect the PPIase activity of GST-FKBP4 or GST-PPIA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service