Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4064

Sigma-Aldrich

Ac-Abu-Tle-Leu-Gln-AFC

≥95% (HPLC)

Synonym(s):

Ac-Abu-Tle-LQ-AFC (Abu=2-Aminobutyrate, Tle=t-Butylglycine, AFC = 4-trifluoromethylcoumaryl-7-amide), Ac-QS1-AFC, Acetyl-L-α-aminobutyroyl-L-tert-leucyl-L-leucyl-L-glutamine-α-(4-trifluoromethylcoumaryl-7-amide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H46F3N5O8
Molecular Weight:
709.75
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

CC(N[C@H](C(N[C@H](C(C[C@H](C(N[C@H](C(NC1=CC2=C(C=C1)C(C(F)(F)F)=CC(O2)=O)=O)CCC(N)=O)=O)CC(C)C)=O)C(C)(C)C)=O)CC)=O

InChI

1S/C34H46F3N5O8/c1-8-23(39-18(4)43)31(48)42-29(33(5,6)7)25(44)14-19(13-17(2)3)30(47)41-24(11-12-27(38)45)32(49)40-20-9-10-21-22(34(35,36)37)16-28(46)50-26(21)15-20/h9-10,15-17,19,23-24,29H,8,11-14H2,1-7H3,(H2,38,45)(H,39,43)(H,40,49)(H,41,47)(H,42,48)/t19

InChI key

VWVFVJSQWAXHGY-RYRALHHHSA-N

Biochem/physiol Actions

Kinetically preferred fluorogenic substrate for detecting SARS-CoV and SARS-CoV-2 main proteases (Mpro; 3CLpro) activity (ex 380+/-20 nm, em 500+/-20 nm).
Ac-QS1-AFC or Ac-Abu-Tle-Leu-Gln-AFC (Abu = Aminobutyrate; Tle = t-Butylglycine; AFC= 7-amino-4-trifluoromethyl coumarin) is a kinetically preferred fluorogenic substrate for the detection of SARS-CoV and SARS-CoV-2 main protease (Mpro; 3CLpro) activity. Mpro cleavage results in the release of the C-terminal AFC and increased fluorescence (ex 380+/-20 nm, em 500+/-20 nm).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service