Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4077

Simeprevir

new

≥98% (HPLC)

Synonym(s):

(2R,3aR,10Z,11aS,12aR,14aR)-N-(cyclopropylsulfonyl)-2,3,3a,4,5,6,7,8,9,11a,12,13,14,14a-tetradecahydro-2-[[7-methoxy-8-methyl-2-[4-(1-methylethyl)-2-thiazolyl]-4-quinolinyl]oxy]-5-methyl-4,14-dioxo-cyclopenta[c]cyclopropa[g][1,6]diazacyclotetradecine-12a(1H)-carboxamide, TMC 435, TMC-435, TMC435, TMC 435350, TMC-435350, TMC435350

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H47N5O7S2
CAS Number:
Molecular Weight:
749.94
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(NC(=O)[C@]32NC(=O)[C@H]4[C@@H](C[C@@H](C4)Oc5c6c(nc(c5)c7[s]cc(n7)C(C)C)c(c(cc6)OC)C)C(=O)N(CCCC\C=C/[C@@H]3C2)C)C1CC1

InChI

1S/C38H47N5O7S2/c1-21(2)30-20-51-35(40-30)29-18-32(26-13-14-31(49-5)22(3)33(26)39-29)50-24-16-27-28(17-24)36(45)43(4)15-9-7-6-8-10-23-19-38(23,41-34(27)44)37(46)42-52(47,48)25-11-12-25/h8,10,13-14,18,20-21,23-25,27-28H,6-7,9,11-12,15-17,19H2,1-5H3,(H,41,4

InChI key

JTZZSQYMACOLNN-VDWJNHBNSA-N

Biochem/physiol Actions

Orally active, potent and selective hepatitis C virus (HCV) NS3 protease inhibitor.

Simeprevir (TMC435; TMC435350) is an orally active, potent and selective hepatitis C virus (HCV) NS3/4A serine protease inhibitor (Ki = 0.5/0.4 nM using genotype 1a (H77)/1b (con1b) protease domain) that effectively blocks HCV replication (EC50/EC90 = 8/24 nM post 72h treatment using Huh7-Luc HCV genotype 1b replicon reporter cell line), while exhibiting no inhibitory potency against a panel of non-HCV DNA and RNA viruses even at concentrations >10 µM.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service