Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4101

TMP269

new

≥98% (HPLC)

Synonym(s):

N-((4-(4-Phenylthiazol-2-yl)tetrahydro-2H-pyran-4-yl)methyl)-3-(5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl)benzamide, TMP 269, TMP-269

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21F3N4O3S
CAS Number:
Molecular Weight:
514.52
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to yellow-orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent and type IIa-selective histone deacetylases (HDAC4/5/7/9) inhibitor in vitro and in vivo.



TMP269 is a potent and type IIa-selective histone deacetylases (HDACs) inhibitor (HDAC4/5/7/9 Ki = 126/80/36/19 nM, IC50 = 157/97/43/23 nM; HDAC6/8 IC50 = 8.2/4.2 μM, HADC1/2/3/10/11 IC50 >100 μM) with an active site zinc-targeting trifluoromethyloxadiazole (TFMO). TMP269 inhibits the growth of AsPC-1 by upregulating FOXO3a expression and displays in vivo therapeutic efficacy in animal models of acute kidney injury (50 mg/kg via i.p. in mice) and cerebral ischemia/reperfusion injury (1-16 mg/kg via i.p. in rats).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service