Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4107

PKM2 inhibitor compound 3k

≥98% (HPLC)

Synonym(s):

Compound 3K, 3-Methyl-1,4-dioxo-1,4-dihydronaphthalen-2-yl)methyl piperidine-1-carbodithioate, C3K, C3k, Compound 3k, MDK 4882, MDK-4882, MDK4882, PKM2-IN-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO2S2
CAS Number:
Molecular Weight:
345.48
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

S(CC2=C(C(=O)c3c(cccc3)C2=O)C)C(=S)N1CCCCC1

InChI

1S/C18H19NO2S2/c1-12-15(11-23-18(22)19-9-5-2-6-10-19)17(21)14-8-4-3-7-13(14)16(12)20/h3-4,7-8H,2,5-6,9-11H2,1H3

InChI key

STAFOGVMELKGRI-UHFFFAOYSA-N

Biochem/physiol Actions

Pyruvate kinase M2 (PKM2) inhibitor superior to shikonin.
Compound 3k (C3k) is a pyruvate kinase M2 (PKM2) inhibitor with 3-fold higher potency than shikonin (IC50 = 2.95 and 8.82 μM, respectively, by in vitro PKM2 LDH coupled assay). C3k exhibits potent cancer cell-selective antiproliferative activity in high PKM2-expressing cancer cultures (HCT116/HeLA/H1299 IC50 = 0.18/0.29/1.56 μM with C3k, 1.84/2.45/1.88 μM with shikonin) with much less cytotoxicity toward normal BEAS-2B human bronchial epithelial cells (IC50 = 18.46 μM; IC50 = 5.36 μM with shikonin).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service