Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

US115250

3 Chamber Reagent Container for Midas®, 150 mL

new

For use with Midas® III-Plus Automated Stainer

Sign Into View Organizational & Contract Pricing


About This Item

packaging

1 ea of

compatibility

for use with Midas® III-Plus Automated Stainer

shipped in

ambient

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service