Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB1032

Sigma-Aldrich

Anti-ULBP1 Antibody, clone 6B17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

UL16-binding protein 1;ALCAN-beta;NKG2D ligand 1;N2DL-1;NKG2DL1;Retinoic acid early transcript 1I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6B17, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 28.0 kDa
observed mol wt ~25 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

chimpanzee

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 6B17 is a Rabbit recombinant monoclonal antibody that specifically detects UL16-binding protein 1. It targets an epitope within 18 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the N-terminal region of human UL16-binding protein 1.

Application

Quality Control Testing

Evaluated by Western Blotting with human recombinant ULBP1.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected human recombinant ULBP1.

Tested applications


Affinity Binding Assay: A representative lot of this antibody bound ULBP1 with a KD of 1.0 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ULBP1in K562 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

UL16-binding protein 1 (UniProt: Q9BZM6; also known as ALCAN-beta, NKG2D ligand 1, N2DL-1, NKG2DL1, Retinoic acid early transcript 1I) is encoded by the ULBP1 (also known as N2DL1, RAET1I) gene (Gene ID: 80329) in human. The UL16-binding proteins are a novel family of MHC class I-related, GPI-anchored, cell surface proteins that lack an 3 domain, and do not associate with β2-microglobulin. ULBP1 was first identified based on its ability to bind to the human CMV (HCMV) glycoprotein UL16. It is synthesized with a signal peptide (aa 1-25) and a propeptide (aa 217-244) that are subsequently cleaved off to produce the mature protein (aa 26-216). It is expressed in T-cells, B-cells, erythroleukemia cell lines and in a wide range of tissues. It is also found in fetal heart, brain, lung, and liver. It binds and activates the natural killer group 2 member D (NKG2D) receptor and mediates natural killer cell cytotoxicity. In CMV-infected cells, it interacts with the viral glycoprotein UL16 and this interaction causes ULBP1 retention in the endoplasmic reticulum and cis-Golgi and prevents binding to and activation of NKG2D, which provides CMV with an immune evasion mechanism. ULBP1 expressed on the tumor cell surface can also bind to the NKG2D receptor presenting on NK cells and CD8+ T and T cells. Patients with NKG2D expression in gastric tumor exhibit significantly longer overall survival compared to those without NKG2D expression in tumors. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Kamei, R., et al. (2018). Oncology Lett. 15(1); 747-754; Sutherland, CL., et al. (2002). J. Immunol. 168(2); 671-679).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service