Skip to Content
MilliporeSigma
All Photos(14)

Key Documents

ZRB1061

Sigma-Aldrich

Anti-MLL/HRX CT Antibody, clone 5C15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5C15, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~431.76 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5C15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Histone-lysine N-methyltransferase 2A (MLL/HRX). It targets an epitope within 21 amino acids from the MLL cleavage product C180 region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the p180 C-terminal subunit of human Histone-lysine N-methyltransferase 2A (MLL/HRX).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected MLL/HRX in human kidney tissue sections.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound MLL peptides with a KD of 1.2 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from representative lot detected MLL/HRX CT in HeLa and A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MLL/HRX CT, clone 5C15 ZooMAb®, Cat. No. ZRB1061, is a recombinant Rabbit monoclonal antibody that specifically targets MLL/HRX and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Immunohistochemistry (Paraffin).

Target description

Histone-lysine N-methyltransferase 2A (UniProt: Q03164; also known as EC:2.1.1.354, Lysine N-methyltransferase 2A, ALL-1, CXXC-type zinc finger protein 7, Myeloid/lymphoid or mixed-lineage leukemia, Myeloid/lymphoid or mixed-lineage leukemia protein 1, MLL, Trithorax-like protein, Zinc finger protein HRX) is encoded by the KMT2A (also known as ALL1, CXXC7, HRX, HTRX, MLL, MLL1, TRX1) gene (Gene ID: 4297) in human. MLL is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is expressed in heart, lung, brain, and in T and B lymphocytes. Catalytic subunit of the MLL complex mediates both methylation of lysine 4 of histone H3 (H3K4me) complex and acetylation of lysine 16 of histone H4 (H4K16ac). By itself it has weak methyltransferase activity and requires another component of the MLL complex to obtain full methyltransferase activity. MLL does not display any activity towards histone H3 phosphorylated on threonine 3 and has less activity towards histone H3 dimethylated on arginine 8 and lysine 9. However, it displays high activity towards histone H3 acetylated on lysine 9. MLL is normally cleaved into a N-terminal p320 and a C-terminal p180 cleavage product. This cleavage is brought about by taspase 1 (TASP1) at cleavage site 1 (aa 2666-2667) and cleavage site 2 (aa 2718-2719). The p320 and p180 units can heterodimerize via SET and FYRC domains. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service