Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

ZRB1740

Sigma-Aldrich

Anti-LRRFIP2 Antibody, clone 1B22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Leucine-rich repeat flightless-interacting protein 2;LRR FLII-interacting protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 82.17 kDa
observed mol wt ~79 kDa

purified by

using Protein A

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LRRFIP2(9209)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B22 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Leucine-rich repeat flightless-interacting protein 2 (LRRFIP2). It targets an epitope within 24 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human Leucine-rich repeat flightless-interacting protein 2 (LRRFIP2).

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LRRFIP2 in A431 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LRRFIP2 in lysate from NIH3T3 cell lysates and recombinant fragment of Human LRRFIP2.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LRRFIP2 in A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound LRRFIP2 peptide with a KD of 5.4 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): A representative lot of this antibody detected recombinant fragment of Human LRRFIP2 in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Leucine-rich repeat flightless-interacting protein 2 (UniProt: Q9Y608; also known as LRR FLII-interacting protein 2, LRRFIP2) is encoded by the LRRFIP2 gene (Gene ID: 9209) in human. LRRFIP2 is a myeloid differentiation factor 88 (MYD88)-interacting protein with a positive regulatory function in toll-like receptor signaling. It is widely expressed with highest levels observed in heart and skeletal muscle. Five isoforms of LRRFIP2 have been described that are produced by alternative splicing. Isoform 1 is long isoform and other isoforms are considered as short isoforms. It interacts with Dvl3 to activate the canonical Wnt signaling pathway and increases β-catenin/ LEF/TCF-dependent transcriptional activity. Its Dvl3-binding region is localized in amino acids 1-370. Both the amino and carboxyl termini of LRRFIP2 are essential for its function. The amino terminus is responsible for interaction with the central region of Dvl3, which contains the PDZ and DEP domains. Mutant form of LRRFIP2 that contains only the amino terminal acts as a dominant negative form. In resting cells, it shows weak interaction with MYD88. However, following LPS-stimulation, the interaction with MYD is rapidly increased and it returns to basal levels after about six hours of stimulation. Serine 190 and 202 of LRRFIP2 are phosphorylated in response to LPS stimulation. Its interaction with MYD is regulated by LPS-induced phosphorylation of serine 202. LRRFIP2 is also reported to negatively regulate NLRP3 inflammasome activation in macrophages by promoting Flightless-I-mediated caspase-1 inhibition. It binds to NLRP3 via its N-terminal upon NLRP3 inflammasome activation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Jin, J., et al. (2013). Nat. Commun. 4; Article 2075; Dai, P., et al. (2009). J. Immunol. 182(6); 3450-3460; Liu, J., et al. (2005). Proc. Natl. Acad. Sci. USA. 102(6); 1927-1932).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service