Skip to Content
MilliporeSigma
All Photos(9)

Key Documents

ZRB2781

Sigma-Aldrich

Anti-Podocalyxin Antibody, clone 1E13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

GCTM-2 antigen, Gp200, PC, PCLP-1, Podocalyxin-like protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E13, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58.64 kDa
observed mol wt ~170 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PODXL(5420)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E13 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Podocalyxin (PODXL). It targets an epitope within 25 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal half of human Podocalyxin.

Application

Quality Control Testing

Evaluated by Western Blotting in Human kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Podocalyxin in Human kidney tissue lysate.

Tested Applications

Flow Cytometry Analysis: 1 µg from a representative lot detected Podocalyxin in one million NTERA-2 cells.

Affinity Binding Assay: A representative lot of this antibody bound Podocalyxin peptide with a KD of 1.3 x 10-6 in an affinity binding assay.

Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected Podocalyxin in Human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Podocalyxin in Human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Podocalyxin (UniProt: O00592; also known as GCTM-2 antigen, Gp200, Podocalyxin-like protein 1, PC, PCLP-1) is encoded by the PODXL (also known as PCLP, PCLP1) gene (Gene ID: 5420) in human. Podocalyxin is a single-pass type I membrane glycoprotein, which is synthesized with a signal peptide (aa 1-22) that is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 23-461), a transmembrane domain (aa 462-482), and a cytoplasmic domain (aa 483-558). Its expression is observed in glomerular epithelium cells (podocytes). It exists in a monomeric form when associated with the membrane raft, but oligomerizes when integrated in the apical membrane. Podocalyxin functions as an anti-adhesive molecule that maintains an open filtration pathway between neighboring foot processes in the podocyte by charge repulsion. It can also act as a pro-adhesive molecule, enhancing the adherence of cells to immobilized ligands, increasing the rate of migration and cell-cell contacts in an integrin-dependent manner. It is reported to induce the formation of apical actin-dependent microvilli and is involved in the formation of a preapical plasma membrane subdomain to set up initial epithelial polarization and the apical lumen formation during renal tubulogenesis. It is reported to play a role in cancer development and aggressiveness by inducing cell migration and invasion through its interaction with the actin-binding protein EZR. It affects EZR-dependent signaling events, leading to increased activities of the MAPK and PI3K pathways in cancer cells. Two isoforms of Podocalyxin have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Nielsen, JS., and McNagny, KM (2009). J. Am. Soc. Nephrol. 20(8); 1669-1676; Sizemore, S., et al. (2007). Cancer Res. 67(13); 6183-6191).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service