Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB3147

Anti-IFT88 Antibody, clone 2C11 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Intraflagellar transport protein 88 homolog, Recessive polycystic kidney disease protein Tg737 homolog, TPR repeat protein 10, Tetratricopeptide repeat protein 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2C11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 93.19 kDa
observed mol wt ~93 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

chimpanzee

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... IFT88(8100)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2C11 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects IFT88. It targets an epitope within 23 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal region of human IFT88.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected IFT88 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected IFT88 in lysates from Jurkat cells and mouse testis tissue.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IFT88 in human testis tissue sections.

Immunofluorescence Analysis: A 1:00 dilution from a representative lot detected IFT88 in human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound IFT88 peptide with at least one hundred thousand-fold (100,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Intraflagellar transport protein 88 homolog (UniProt: Q13099; also known as Polaris, Recessive polycystic kidney disease protein Tg737 homolog, Tetratricopeptide repeat protein 10, TPR repeat protein 10) is encoded by the IFT88 (also known as TG737, TTC10 ) gene (Gene ID: 8100) in human. The primary cilium is a microtubule-based mechano and chemo-sensory organelle that coordinates an array of cellular pathways during development and in tissue homeostasis, including Hedgehog (Hh), PDGFR and Wnt signaling. IFT88 is primarily localized at the basal body of primary cilia and within the cilia. Primary cilium is assembled and maintained via a process termed intraflagellar transport (IFT) in mammalian cells during growth arrest. IFT88 is a subunit of the IFT particle complex B required for the assembly of functional IFT. It is widely expressed in various tissues, particularly heart, brain, liver, lung, kidney, skeletal muscle and pancreas. Three isoforms of IFT88 have been described that are produced by alternative splicing. IFT88 protein has 9 Tetratricopeptide repeats (TPR) which are functional in protein-protein interactions. Defects in primary cilium assembly lead to severe developmental diseases and disorders referred to as ciliopathies. Polycystic kidney disease (PKD) is one of the first diseases identified to be linked to dysfunctional primary cilia. PKD was originally identified in the Oak Ridge Polycystic Kidney (ORPK) mouse harboring mutated IFT88 gene (Ift88orpk or Ift88Tg737NRpw). Studies have shown that loss of function of IFT88/primary cilia does not promote thyroid cancer cell proliferation, migration, and invasion but, IFT88 may play a role in metabolic reprogramming in thyroid cancer cells. IFT88 is commonly used as a primary cilium marker. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lee, J., et al. (2018). Oncogene. 37;4455-4474; Clement, A C., et al. (2009). Methods in Cell Biology. 94;181-197).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service