Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72691

Supelco

Pristanic acid

new

mixture of isomers, analytical standard

Synonym(s):

2,6,10,14-Tetramethylpentadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H38O2
CAS Number:
Molecular Weight:
298.50
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Assay

≥95.0% (mixture of isomers)

quality

mixture of isomers

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC(=O)C(CCCC(CCCC(CCCC(C)C)C)C)C

InChI key

PAHGJZDQXIOYTH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service