Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93056

Supelco

Alternariol solution

20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,7,9-Trihydroxy-1-methyl-6H-dibenzo[b,d]pyran-6-one solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O5
CAS Number:
Molecular Weight:
258.23
MDL number:
UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in acetonitrile (nominal concentration)

technique(s)

HPLC: suitable
LC/MS: suitable
liquid chromatography (LC): suitable
mass spectrometry (MS): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[o]1c2c(c3c([c]1=O)c(cc(c3)O)O)c(cc(c2)O)C

InChI

1S/C14H10O5/c1-6-2-7(15)5-11-12(6)9-3-8(16)4-10(17)13(9)14(18)19-11/h2-5,15-17H,1H3

InChI key

CEBXXEKPIIDJHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.Certified content incl. uncertainty and expiry date are given on the certificate.

Download your certificate at: http://www.sigma-aldrich.com.

We also offer a CRM solution of isotope labelled Alternariol. Please find all our CRMs of Alternaria toxins here.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service