Skip to Content
MilliporeSigma

1026412

USP

3-Aminosalicylic Acid

new

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Aminosalicylic acid, 3-Amino-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
H2NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
2090427
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

pharmaceutical primary standard

Agency

certified by the USP

manufacturer/tradename

USP

mp

240 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Nc1cccc(C(O)=O)c1O

InChI

1S/C7H7NO3/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,9H,8H2,(H,10,11)

InChI key

IQGMRVWUTCYCST-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service