Skip to Content
MilliporeSigma

1086414

USP

Calcium citrate tetrahydrate

new

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,2,3-Propanetricarboxylic acid, 2-hydroxy-, calcium salt (2:3), tetrahydrate, Tricalcium dicitrate tetrahydrate

Sign Into View Organizational & Contract Pricing

About This Item

Linear Formula:
[O2CCH2C(OH)(CO2)CH2CO2]2Ca3 · 4H2O
CAS Number:
Molecular Weight:
570.49
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

pharmaceutical primary standard

Agency

USP

manufacturer/tradename

USP

application(s)

(Pharmaceutical small molecule)

format

neat

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].OC1(CC(=O)O[Ca]OC(=O)C1)C(=O)O[Ca]OC(=O)C2(O)CC(=O)O[Ca]OC(=O)C2

InChI

1S/2C6H8O7.3Ca.4H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;;3*+2;;;;/p-6

InChI key

LNIZKKFWMDARJV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service