Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1A00190

USP

QUETIAPINE DESETHOXY

new

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Quetiapine Desethoxy, (2-[4-(dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3OS
CAS Number:
Molecular Weight:
339.45
MDL number:
UNSPSC Code:
12352200

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

manufacturer/tradename

USP

application(s)

pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[s]1c2c(nc(c4c1cccc4)N3CCN(CC3)CCO)cccc2

InChI

1S/C19H21N3OS/c23-14-13-21-9-11-22(12-10-21)19-15-5-1-3-7-17(15)24-18-8-4-2-6-16(18)20-19/h1-8,23H,9-14H2

InChI key

OFLMIXVKBNAUIB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

USP issued SDS can be found here
Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service