MilliporeSigma
BR781346
BRAND® 384-well PCR plate
new
PP, 0.03 ml, full-skirted, transparent, BIO-CERT® PCR QUALITY, 50EA
About This Item
Code UNSPSC :
41121800
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Matériaux
transparent polypropylene plate
Stérilité
non-sterile
Caractéristiques
DNA free
RNase and DNase free
pyrogen free
skirt
Conditionnement
pack of 50 ea
Fabricant/nom de marque
BRAND 781346
Taille
384 well
Puits
384
Volume de travail
30 μL
Adéquation
suitable for PCR
Type de liaison
non-treated surface
Description générale
BRAND® 384-well PCR plates are an economical solution for automatic high-throughput analyses. The thin walls allow fast transmission of the temperature specified by the cycler, thus reducing cycle times. The stable design of the plates makes them ideal for automation, as they can be gripped by robot systems without twisting. To minimize evaporation with small sample volumes to 2 µl and prevent contamination, seal the plates with the appropriate sealing film. The cleanroom quality of the PCR consumables, including 384-well PCR plates from BRAND® guarantees reliable results.
Caractéristiques et avantages
  • For applications with automation and high-throughput analyses
  • Rapid heat transfer through thin vessel walls
  • Can be sealed reliably with self-adhesive sealing films
  • BIO-CERT® PCR QUALITY: Free of DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Informations légales
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG