SNAP2IHC
SNAP id® 2.0 Protein Detection System
Single IHC
About This Item
Code UNSPSC :
41116010
eCl@ss :
42029053
Nomenclature NACRES :
NB.23
Produits recommandés
Product Name
SNAP i.d. 2.0 Protein Detection System-Single IHC,
Gamme de produits
SNAP id®
Technique(s)
immunohistochemistry: suitable (paraffin)
immunohistochemistry: suitable (tissue)
immunohistochemistry: suitable
Description générale
This system includes the SNAP id® 2.0 Base, IHC frame and incubation cover, assembly fixture, vacuum tubing, and a Quick Start Guide.
Application
The SNAP id® 2.0 System for IHC introduces a streamlined, medium through-put IHC capability to our SNAP id® 2.0 family. The frame and slide holders enable you to process up to 12 tissue slides, while our unique vacuum technology evenly removes
Autres remarques
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Informations légales
SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany