Principaux documents
TRCSINTFILTIL
Introduction to Filtration (On-site Training)
About This Item
Code UNSPSC :
86132100
Description générale
Using a combination of theory and hands-on instruction, you will learn the key procedures of filter integrity testing, how to read printouts from automatic filter-integrity testers and troubleshoot filter-integrity test processes. This two-day course covers all current filter integrity tests, including bubble point, diffusive flow and water-flow integrity testing that apply to all liquid and gas membrane filtration applications.